4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic Acid, 4-(2-(2-Amino-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzoic acid, 2-Amino-4,7-dihydro-5-[2-[4-(carboxy)phenyl]ethyl]-4-oxo-3H-pyrrolo[2,3-d]pyrimidine

A2618-1G A2618-5G
TCA2618-1GEA 64.78 CAD
TCA2618-1G TCA2618-5G
4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid hydrate ≥98.0% (by HPLC, titration analysis)
4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid hydrate

CAS Number: 137281-39-1
MDL Number: MFCD11040439
Molecular Formula: C15H14N4O3
Molecular Weight: 298.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 2-Amino-4,7-dihydro-5-[2-[4-(carboxy)phenyl]ethyl]-4-oxo-3H-pyrrolo[2,3-d]pyrimidine

Formula: C₁₅H₁₄N₄O₃
MW: 298.30 g/mol
MDL Number: MFCD11040439
CAS Number: 137281-39-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02e257393d45ab21c9fe8ac0570648fe2fb6b9e3&vid=f474c78b3c4a9b07326760510db1c0e1096616bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR