4-(4-Aminophenoxy)-N-methylpicolinamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-(4-Aminophenoxy)-N-methylpicolinamide, 4-(4-Aminophenoxy)-N-methyl-2-pyridinecarboxamide

A2617-1G A2617-5G
TCA2617-1GEA 45.82 CAD
TCA2617-1G TCA2617-5G
4-(4-Aminophenoxy)-N-methylpicolinamide ≥98.0% (by HPLC, titration analysis)
4-(4-Aminophenoxy)-N-methylpicolinamide

CAS Number: 284462-37-9
MDL Number: MFCD08692047
Molecular Formula: C13H13N3O2
Molecular Weight: 243.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 118


Other Product Names / Synonyms: 4-(4-Aminophenoxy)-N-methylpicolinamide

Formula: C₁₃H₁₃N₃O₂
MW: 243.2612 g/mol
Melting Pt: 118 °C
MDL Number: MFCD08692047
CAS Number: 284462-37-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1ec002fae3680708707ba1ea074e113596cf114&vid=e1ec002fae3680708707ba1ea074e113596cf114&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR