4-(4-Aminophenyl)morpholin-3-one ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2614-1G A2614-5G
TCA2614-1GEA 26.86 CAD
TCA2614-1G TCA2614-5G
4-(4-Aminophenyl)morpholin-3-one ≥98.0% (by GC, titration analysis)
4-(4-Aminophenyl)morpholin-3-one

CAS Number: 438056-69-0
MDL Number: MFCD08236742
Molecular Formula: C10H12N2O2
Molecular Weight: 192.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 173


Other Product Names / Synonyms: 4-(3-Oxomorpholino)aniline

Formula: C₁₀H₁₂N₂O₂
Melting Pt: 173 °C
MDL Number: MFCD08236742
CAS Number: 438056-69-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec91f1cd7fc7eedb5eff8b4c26f9c49101637fd2&vid=ec91f1cd7fc7eedb5eff8b4c26f9c49101637fd2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR