5-Aminobenzimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2605-1G A2605-5G
TCA2605-1GEA 152 CAD
TCA2605-1G TCA2605-5G
5-Aminobenzimidazole ≥98.0% (by GC, titration analysis)
5-Aminobenzimidazole

CAS Number: 934-22-5
MDL Number: MFCD00465258
Molecular Formula: C7H7N3
Molecular Weight: 133.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 222
Melting point (°C): 163
Lambda max.: 300 nm (EtOH)

Formula: C₇H₇N₃
Boiling Pt: 222 °C
Melting Pt: 163 °C
MDL Number: MFCD00465258
CAS Number: 934-22-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18acb7f4b92991326089fed3c46d710062b1db48&vid=324b819dda7750bf72a5a77f36292a2abd9786f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR