Methyl-3-aminopropanoate hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: beta-Alanine methyl ester hydrochloride, H-β-Ala-OMe·HCl, β-Alanine methyl ester hydrochloride, 3-Aminopropionic acid Methyl Ester Hydrochloride, 3-Aminopropionic Acid Methyl Ester Hydrochloride, Methyl 3-aminopropanoate hydrochloride, Methyl 3-Aminopropionate Hydrochloride, H-β-Ala-OMe.HCl

A2603-25G A2603-5G
TCA2603-25GEA 246.48 CAD
TCA2603-25G TCA2603-5G
Methyl-3-aminopropanoate hydrochloride ≥98.0%
Methyl-3-aminopropanoate hydrochloride

CAS Number: 3196-73-4
MDL Number: MFCD00039060
Molecular Formula: C4H9NO2
Molecular Weight: 139.58
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 108
Storage Temperature: 0-10°C


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: Methyl 3-Aminopropionate Hydrochloride, 3-Aminopropionic Acid Methyl Ester Hydrochloride

Formula: C4H10ClNO2
MW: 139.6 g/mol
Melting Pt: 108 °C
MDL Number: MFCD00039060
CAS Number: 3196-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4822498397cae02a389af288ca27f760614242e6&vid=4822498397cae02a389af288ca27f760614242e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR