3-Amino-1-propanesulfonic acid ≥98.0%

Supplier: TCI America

Synonyms: 3-Aminopropane-1-sulfonic acid, Tramiprosate, Homotaurine, 3-Aminopropane-1-sulphonic acid, 3APS

A2602-25G A2602-5G
TCA2602-25GEA 509.64 CAD
TCA2602-25G TCA2602-5G
3-Amino-1-propanesulfonic acid ≥98.0%
3-Amino-1-propanesulfonic acid

CAS Number: 3687-18-1
MDL Number: MFCD00008225
Molecular Formula: C3H9NO3S
Molecular Weight: 139.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 292


Product Categories: Synthetic Organic Chemistry, Sulfonic Acids, Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 3APS, Homotaurine, Tramiprosate

Formula: C₃H₉NO₃S
Melting Pt: 292 °C
MDL Number: MFCD00008225
CAS Number: 3687-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e561e455f5341ae39eb4c541b03204f8eda2530&vid=9e561e455f5341ae39eb4c541b03204f8eda2530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR