Azilsartan ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Ethoxy-1-((2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-benzo[d]imidazole-7-carboxylic acid

A2601-1G A2601-200MG
TCA2601-1GEA 546.68 CAD
TCA2601-1G TCA2601-200MG
Azilsartan ≥98.0% (by HPLC, titration analysis)
Azilsartan

CAS Number: 147403-03-0
MDL Number: MFCD00916395
Molecular Formula: C25H20N4O5
Molecular Weight: 456.46
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 188
Storage Temperature: 0-10°C

Formula: C₂₅H₂₀N₄O₅
MW: 456.45 g/mol
Melting Pt: 188 °C
MDL Number: MFCD20278186
CAS Number: 147403-03-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c54d989f01ec65916d80293864d610c478f8a42&vid=9c54d989f01ec65916d80293864d610c478f8a42&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR