2-Amino-N-methylbenzamide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2597-1G A2597-5G
TCA2597-1GEA 95.91 CAD
TCA2597-1G TCA2597-5G
2-Amino-N-methylbenzamide ≥98.0% (by GC, titration analysis)
2-Amino-N-methylbenzamide

CAS Number: 4141-08-6
MDL Number: MFCD00060602
Molecular Formula: C8H10N2O
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 79


Other Product Names / Synonyms: 2-Aminobenzoylmethylamide

Formula: C₈H₁₀N₂O
MW: 150.1778 g/mol
Melting Pt: 79 °C
MDL Number: MFCD00060602
CAS Number: 4141-08-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22ee83a9bb9fcbc5aa76a71dcda1612c29011a74&vid=22ee83a9bb9fcbc5aa76a71dcda1612c29011a74&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR