Isoquinoline-4-amine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2595-1G A2595-5G
TCA2595-1GEA 72.68 CAD
TCA2595-1G TCA2595-5G
Isoquinoline-4-amine ≥98.0% (by GC, titration analysis)
Isoquinoline-4-amine

CAS Number: 23687-25-4
MDL Number: MFCD00034752
Molecular Formula: C9H8N2
Molecular Weight: 144.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 110

Formula: C₉H₈N₂
Melting Pt: 110 °C
MDL Number: MFCD00034752
CAS Number: 23687-25-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cddf993fa539c1fbcc523aa785e40b78995dab9&vid=3cddf993fa539c1fbcc523aa785e40b78995dab9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR