4-(trans-4-Amylcyclohexyl)benzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2593-25G A2593-5G
TCA2593-25GEA 161.95 CAD
TCA2593-25G TCA2593-5G
4-(trans-4-Amylcyclohexyl)benzoic acid ≥98.0% (by GC, titration analysis)
4-(trans-4-Amylcyclohexyl)benzoic acid

CAS Number: 65355-30-8
MDL Number: MFCD06208360
Molecular Formula: C18H26O2
Molecular Weight: 274.40
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 181


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-(trans-4-Pentylcyclohexyl)benzoic Acid

Formula: C₁₈H₂₆O₂
Melting Pt: 181 °C
MDL Number: MFCD06208360
CAS Number: 65355-30-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66497f8a3bfe20f5e3f2316863057d7f46d97467&vid=66497f8a3bfe20f5e3f2316863057d7f46d97467&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR