ɑ-Asarone ≥98.0% (by GC)

Supplier: TCI America

A2590-1G A2590-5G
TCA2590-1GEA 60.04 CAD
TCA2590-1G TCA2590-5G
ɑ-Asarone ≥98.0% (by GC)
alpha-Asarone

CAS Number: 2883-98-9
MDL Number: MFCD00064457
Molecular Formula: C12H16O3
Molecular Weight: 208.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 298
Melting point (°C): 61
Lambda max.: 312 nm (EtOH)


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use
Other Product Names / Synonyms: trans-1,2,4-Trimethoxy-5-(1-propenyl)benzene

Formula: C₁₂H₁₆O₃
Boiling Pt: 298 °C
Melting Pt: 61 °C
MDL Number: MFCD00064457
CAS Number: 2883-98-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d9bb08ac9bf9e2164a8dfbdb9a35986f78e3dbc&vid=1d9bb08ac9bf9e2164a8dfbdb9a35986f78e3dbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR