(S)-Isopropyl-2-aminopropanoate hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A2588-25G
A2588-5G
TCA2588-25GEA
154.84
CAD
TCA2588-25G
TCA2588-5G
(S)-Isopropyl-2-aminopropanoate hydrochloride ≥98.0% (by titrimetric analysis)
(S)-Isopropyl-2-aminopropanoate hydrochloride
CAS Number: 62062-65-1
MDL Number: MFCD08059709
Molecular Formula: C6H13NO2
Molecular Weight: 167.63
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 85
Specific rotation [a]20/D: 2 deg (C=2, MeOH)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: (S)-2-Aminopropionic Acid Isopropyl Ester Hydrochloride, Isopropyl (S)-2-Aminopropionate Hydrochloride
Formula:
C6H14ClNO2 Melting Pt: 85 °C |
MDL Number:
MFCD08059709 CAS Number: 62062-65-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...