2-Amino-3-bromo-5-methylbenzoic acid ≥97.0%

Supplier: TCI America

A2569-1G A2569-5G
TCA2569-1GEA 63.2 CAD
TCA2569-1G TCA2569-5G
2-Amino-3-bromo-5-methylbenzoic acid ≥97.0%
2-Amino-3-bromo-5-methylbenzoic acid

CAS Number: 13091-43-5
MDL Number: MFCD00051705
Molecular Formula: C8H8BrNO2
Molecular Weight: 230.06
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 206


Other Product Names / Synonyms: 3-Bromo-5-methylanthranilic Acid, 6-Amino-5-bromo-m-toluic Acid

Formula: C₈H₈BrNO₂
MW: 230.06
Melting Pt: 206 °C
MDL Number: MFCD00051705
CAS Number: 13091-43-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6549a574a2c405ff3a3d957d6fd61c830a025996&vid=6549a574a2c405ff3a3d957d6fd61c830a025996&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR