trans-4-[4-(4-Pentylcyclohexyl)phenyl]benzonitrile ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4'-(trans-4-Pentylcyclohexyl)-[1,1'-biphenyl]-4-carbonitrile
A2568-1G
A2568-5G
TCA2568-1GEA
104.12
CAD
TCA2568-1G
TCA2568-5G
trans-4-[4-(4-Pentylcyclohexyl)phenyl]benzonitrile ≥98.0% (by GC)
trans-4-[4-(4-Pentylcyclohexyl)phenyl]benzonitrile
CAS Number: 68065-81-6
MDL Number: MFCD01076317
Molecular Formula: C24H29N
Molecular Weight: 331.50
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 211
Melting point (°C): 96
Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Formula:
C₂₄H₂₉N MW: 331.50 g/mol Boiling Pt: 211 °C Melting Pt: 96 °C |
MDL Number:
MFCD01076317 CAS Number: 68065-81-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...