2-Amino-5-[4-[2-(5-ethyl-2-pyridyl)ethoxy]benzyl]thiazol-4(5H)-one ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2-imino-4-thiazolidinone, 2-Amino-5-(4-(2-(5-ethylpyridin-2-yl)ethoxy)benzyl)thiazol-4(5H)-one

A2567-25G A2567-5G
TCA2567-25GEA 319 CAD
TCA2567-25G TCA2567-5G
2-Amino-5-[4-[2-(5-ethyl-2-pyridyl)ethoxy]benzyl]thiazol-4(5H)-one ≥98.0% (by HPLC, titration analysis)
2-Amino-5-[4-[2-(5-ethyl-2-pyridyl)ethoxy]benzyl]thiazol-4(5H)-one

CAS Number: 105355-26-8
MDL Number: MFCD07368627
Molecular Formula: C19H21N3O2S
Molecular Weight: 355.46
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 196


Other Product Names / Synonyms: 5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2-imino-4-thiazolidinone

Formula: C₁₉H₂₁N₃O₂S
MW: 355.46 g/mol
Melting Pt: 196 °C
MDL Number: MFCD07368627
CAS Number: 105355-26-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd9ff340f0f0d9a6f64809f534744a886cad9fd8&vid=fd9ff340f0f0d9a6f64809f534744a886cad9fd8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR