1-Amino-2-methylpyridinium iodide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A2558-1G
A2558-5G
TCA2558-1GEA
153.26
CAD
TCA2558-1G
TCA2558-5G
1-Amino-2-methylpyridinium iodide ≥98.0% (by HPLC, titration analysis)
1-Amino-2-methylpyridinium iodide
CAS Number: 7583-90-6
MDL Number: MFCD00955862
Molecular Formula: C6H9IN2
Molecular Weight: 236.06
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 152
Formula:
C₆H₉IN₂ Melting Pt: 152 °C |
MDL Number:
MFCD00955862 CAS Number: 7583-90-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...