4-Amino-2-methoxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Methoxypyridin-4-amine

A2543-1G A2543-5G
TCA2543-1GEA 37.92 CAD
TCA2543-1G TCA2543-5G
4-Amino-2-methoxypyridine ≥98.0% (by GC, titration analysis)
4-Amino-2-methoxypyridine

CAS Number: 20265-39-8
MDL Number: MFCD06738657
Molecular Formula: C6H8N2O
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 90

Formula: C₆H₈N₂O
MW: 124.1400 g/mol
Melting Pt: 90 °C
MDL Number: MFCD06738657
CAS Number: 20265-39-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43abd6a005080b4c2722c0e89aa35ebbc723a5f5&vid=43abd6a005080b4c2722c0e89aa35ebbc723a5f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR