2-Amino-3-iodopyridine ≥98.0%
Supplier: TCI America
A2542-1G
A2542-5G
TCA2542-1GEA
47.87
CAD
TCA2542-1G
TCA2542-5G
2-Amino-3-iodopyridine ≥98.0%
2-Amino-3-iodopyridine
CAS Number: 104830-06-0
MDL Number: MFCD01651885
Molecular Formula: C5H5IN2
Molecular Weight: 220.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 91
Formula:
C₅H₅IN₂ Melting Pt: 91 °C |
MDL Number:
MFCD01651885 CAS Number: 104830-06-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...