3-Amino-5-bromo-2-chloropyridine ≥98.0% (by GC)
Supplier: TCI America
A2540-25G
A2540-5G
TCA2540-25GEA
744.18
CAD
TCA2540-25G
TCA2540-5G
3-Amino-5-bromo-2-chloropyridine ≥98.0% (by GC)
3-Amino-5-bromo-2-chloropyridine
CAS Number: 588729-99-1
MDL Number: MFCD02682092
Molecular Formula: C5H4BrClN2
Molecular Weight: 207.46
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 131
Lambda max.: 314 nm (EtOH)
Formula:
C₅H₄BrClN₂ Melting Pt: 131 °C |
MDL Number:
MFCD02682092 CAS Number: 588729-99-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...