5-Amino-3-bromo-2-chloropyridine ≥95.0% (by GC)

Supplier: TCI America

A2539-1G A2539-5G
TCA2539-1GEA 28.44 CAD
TCA2539-1G TCA2539-5G
5-Amino-3-bromo-2-chloropyridine ≥95.0% (by GC)
5-Amino-3-bromo-2-chloropyridine

CAS Number: 130284-53-6
MDL Number: MFCD03840433
Molecular Formula: C5H4BrClN2
Molecular Weight: 207.46
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 113

Formula: C₅H₄BrClN₂
Melting Pt: 113 °C
MDL Number: MFCD03840433
CAS Number: 130284-53-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74bfb57fac7366a1efd416cc80ee658276ac5d89&vid=74bfb57fac7366a1efd416cc80ee658276ac5d89&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR