2-Amino-4-methylbenzoic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-Amino-p-toluic acid, 4-Methylanthranilic acid, 4-Methylanthranil acid, 2-Amino-4-methylbenzoic Acid

A2538-1G A2538-5G
TCA2538-1GEA 214.88 CAD
TCA2538-1G TCA2538-5G
2-Amino-4-methylbenzoic acid ≥98.0%
2-Amino-4-methylbenzoic acid

CAS Number: 2305-36-4
MDL Number: MFCD00047853
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 171


Other Product Names / Synonyms: 2-Amino-p-toluic Acid, 4-Methylanthranilic Acid

Formula: C₈H₉NO₂
MW: 151.16
Melting Pt: 171 °C
MDL Number: MFCD00047853
CAS Number: 2305-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2846bad94ddd6acba6ffadd966fd5e1773621248&vid=2846bad94ddd6acba6ffadd966fd5e1773621248&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR