1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic acid ≥95.0% (by HPLC)

Supplier: TCI America

A2526-100MG A2526-500MG
TCA2526-100MGEA 281.24 CAD
TCA2526-100MG 76218-746
1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic acid ≥95.0% (by HPLC)
1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic acid

CAS Number: 905954-28-1
Molecular Formula: C15H31NO8
Molecular Weight: 353.41
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal


Product Categories: Bioscience, Crosslinkers, PEGylation Reagents & PEG Spacers
Other Product Names / Synonyms: 21-Amino-4,7,10,13,16,19-hexaoxahenicosanoic Acid, NH2-PEG6-COOH, Amino-PEG6-carboxylate

Formula: C₁₅H₃₁NO₈
MDL Number: MFCD13184943
CAS Number: 905954-28-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2cedb8779246e4d2761615a762ff5abc89e1524&vid=ea3ea52856ac4cc8e0bfe3e12b5d6affa99510c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR