2-Amino-4-(trifluoromethyl)pyrimidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2522-1G A2522-5G
TCA2522-1GEA 74.26 CAD
TCA2522-1G TCA2522-5G
2-Amino-4-(trifluoromethyl)pyrimidine ≥98.0% (by GC, titration analysis)
2-Amino-4-(trifluoromethyl)pyrimidine

CAS Number: 16075-42-6
MDL Number: MFCD00828624
Molecular Formula: C5H4F3N3
Molecular Weight: 163.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 176
Lambda max.: 308 nm (EtOH aq.)

Formula: C₅H₄F₃N₃
Melting Pt: 176 °C
MDL Number: MFCD00828624
CAS Number: 16075-42-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c0e794bacfb319934bf820839e096b964da19fb&vid=8c0e794bacfb319934bf820839e096b964da19fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR