2-Benzylaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Aminodiphenylmethane, 2-Benzylaniline
A2519-25G
A2519-5G
TCA2519-25GEA
375.9
CAD
TCA2519-25G
TCA2519-5G
2-Benzylaniline ≥98.0% (by GC, titration analysis)
2-Benzylaniline
CAS Number: 28059-64-5
MDL Number: MFCD00007750
Molecular Formula: C13H13N
Molecular Weight: 183.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 172
Melting point (°C): 51
Flash Point (°C): 113
Other Product Names / Synonyms: 2-Benzylaniline
Formula:
C₁₃H₁₃N MW: 183.25 g/mol Boiling Pt: 172 °C Melting Pt: 51 °C Flash Pt: 113 °C |
MDL Number:
MFCD00007750 CAS Number: 28059-64-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...