(R)-(+)-ɑ-Amino-γ-butyrolactone hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

Synonyms: D-(+)-Homoserine Lactone Hydrochloride, (R)-(+)-3-Aminotetrahydrofuran-2-one Hydrochloride, (R)-3-Aminodihydrofuran-2(3H)-one hydrochloride

A2512-1G A2512-5G
TCA2512-1GEA 173.48 CAD
TCA2512-1G TCA2512-5G
(R)-(+)-ɑ-Amino-γ-butyrolactone hydrochloride ≥98.0% (by total nitrogen and titration analysis)
(R)-(+)-alpha-Amino-gamma-butyrolactone hydrochloride

CAS Number: 104347-13-9
MDL Number: MFCD00674071
Molecular Formula: C4H7NO2
Molecular Weight: 137.56
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 223
Specific rotation [a]20/D: 29 deg (C=0.2, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(+)-3-Aminotetrahydrofuran-2-one Hydrochloride, D-(+)-Homoserine Lactone Hydrochloride

Formula: C₄H₇NO₂
MW: 137.56 g/mol
Melting Pt: 223 °C
MDL Number: MFCD00674071
CAS Number: 104347-13-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c57fe52bcc0c22e25045598b2893f1386f70af34&vid=6022619ba93c643e924c1a8dc04134ca3b976f4d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR