(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol ≥98.0%

Supplier: TCI America

Synonyms: (S)-2-Amino-3-methyl-1,1-diphenylbutan-1-ol

A2504-1G A2504-5G
TCA2504-1GEA 171.9 CAD
TCA2504-1G TCA2504-5G
(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol ≥98.0%
(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol

CAS Number: 78603-95-9
MDL Number: MFCD01318248
Molecular Formula: C17H21NO
Molecular Weight: 255.36
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 98
Specific rotation [a]20/D: -142 deg (C=1, CHCl3)

Formula: C₁₇H₂₁NO
Melting Pt: 98 °C
MDL Number: MFCD01318248
CAS Number: 78603-95-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2e65e4c76d4c02482f61f25294f529eb9fc126c&vid=f2e65e4c76d4c02482f61f25294f529eb9fc126c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR