(S)-(-)-2-Amino-3,3-dimethyl-1,1-diphenyl-1-butanol ≥98.0% (by HPLC)

Supplier: TCI America

A2503-1G A2503-5G
TCA2503-1GEA 192.76 CAD
TCA2503-1G TCA2503-5G
(S)-(-)-2-Amino-3,3-dimethyl-1,1-diphenyl-1-butanol ≥98.0% (by HPLC)
(S)-(-)-2-Amino-3,3-dimethyl-1,1-diphenyl-1-butanol

CAS Number: 144054-70-6
MDL Number: MFCD03427198
Molecular Formula: C18H23NO
Molecular Weight: 269.39
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 134
Specific rotation [a]20/D: -146 deg (C=0.4, EtOH)

Formula: C₁₈H₂₃NO
MW: 269.39 g/mol
Melting Pt: 134 °C
MDL Number: MFCD03427198
CAS Number: 144054-70-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1689130c40de46f67aebc4c0c84c10aca193d8c&vid=d1689130c40de46f67aebc4c0c84c10aca193d8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR