4-Acetamido-N-(2-aminophenyl)benzamide ≥98.0% (by HPLC)

Supplier: TCI America

A2501-10MG A2501-50MG
TCA2501-10MGEA 101.12 CAD
TCA2501-10MG TCA2501-50MG
4-Acetamido-N-(2-aminophenyl)benzamide ≥98.0% (by HPLC)
4-Acetamido-N-(2-aminophenyl)benzamide

CAS Number: 112522-64-2
MDL Number: MFCD00866266
Molecular Formula: C15H15N3O2
Molecular Weight: 269.30
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 242
Storage Temperature: 0-10°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antitumor Ingredients, Epigenetics Research Reagents
Other Product Names / Synonyms: Acetyldinaline

Formula: C₁₅H₁₅N₃O₂
Melting Pt: 242 °C
MDL Number: MFCD00866266
CAS Number: 112522-64-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14712aab158e54689031802c68c4b5f2ef7e7eb8&vid=14712aab158e54689031802c68c4b5f2ef7e7eb8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR