L-Alanine ethyl ester hydrochloride 98,0

Supplier: TCI America

Synonyms: H-Ala-OEt.HCl, Ethyl (S)-2-Aminopropionate Hydrochloride, L-Ethyl alanate hydrochloride, (S)-2-Aminopropionic Acid Ethyl Ester Hydrochloride, H-Ala-OEt·HCl

A2494-25G A2494-5G
TCA2494-25GEA 139.04 CAD
TCA2494-25G TCA2494-5G
L-Alanine ethyl ester hydrochloride 98,0
L-Alanine ethyl ester hydrochloride

CAS Number: 1115-59-9
MDL Number: MFCD00063662
Molecular Formula: C5H11NO2
Molecular Weight: 153.61
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 79
Specific rotation [a]20/D: 4 deg (C=2.5, H2O)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: (S)-2-Aminopropionic Acid Ethyl Ester Hydrochloride, Ethyl (S)-2-Aminopropionate Hydrochloride, H-Ala-OEt.HCl

Formula: C₅H₁₁NO₂
MW: 153.61 g/mol
Melting Pt: 79 °C
MDL Number: MFCD00063662
CAS Number: 1115-59-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a07d08da040fb0ff885e941062f87fd17468299&vid=8a07d08da040fb0ff885e941062f87fd17468299&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR