Aloe emodin ≥97.0% (by HPLC)

Supplier: TCI America

A2491-100MG A2491-1G
TCA2491-100MGEA 276.03 CAD
TCA2491-100MG TCA2491-1G
Aloe emodin ≥97.0% (by HPLC)
Aloe emodin

CAS Number: 481-72-1
MDL Number: MFCD00017373
Molecular Formula: C15H10O5
Molecular Weight: 270.24
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 222
Storage Temperature: 0-10°C


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Other Pharmaceutical Ingredients, Others (Phytochemicals)
Other Product Names / Synonyms: 1,8-Dihydroxy-3-(hydroxymethyl)anthraquinone

Formula: C₁₅H₁₀O₅
Melting Pt: 222 °C
MDL Number: MFCD00017373
CAS Number: 481-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a5387c09dc82dd97b48fe8e5d4511a6c4e7eaa2&vid=3a5387c09dc82dd97b48fe8e5d4511a6c4e7eaa2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR