Quinoxalin-6-amine ≥98.0%

Supplier: TCI America

Synonyms: 6-Aminoquinoxaline

A2490-1G A2490-5G
TCA2490-1GEA 71.1 CAD
TCA2490-1G TCA2490-5G
Quinoxalin-6-amine ≥98.0%
Quinoxalin-6-amine

CAS Number: 6298-37-9
MDL Number: MFCD00462821
Molecular Formula: C8H7N3
Molecular Weight: 145.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 159

Formula: C₈H₇N₃
MW: 145.1600 g/mol
Melting Pt: 159 °C
MDL Number: MFCD00462821
CAS Number: 6298-37-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ea46fb5f59c6de13d5aaba7996a3c8313a14ce0&vid=2ea46fb5f59c6de13d5aaba7996a3c8313a14ce0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR