3-Amino-9H-pyrido[3,4-b]indole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A2486-100MG A2486-1G
TCA2486-100MGEA 373.51 CAD
TCA2486-100MG TCA2486-1G
3-Amino-9H-pyrido[3,4-b]indole ≥98.0% (by HPLC, titration analysis)
3-Amino-9H-pyrido[3,4-b]indole

CAS Number: 73834-77-2
MDL Number: MFCD00209818
Molecular Formula: C11H9N3
Molecular Weight: 183.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 291
Lambda max.: 299 nm


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 3-Amino-beta-carboline

Formula: C₁₁H₉N₃
Melting Pt: 291 °C
MDL Number: MFCD00209818
CAS Number: 73834-77-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db7ebf72e96bba9363fbfbf6317599e90be31039&vid=db7ebf72e96bba9363fbfbf6317599e90be31039&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR