(Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetic acid ≥98.0%

Supplier: TCI America

Synonyms: (Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetic acid, syn-Thiazoximic Acid

A2478-25G A2478-5G
TCA2478-25GEA 127.35 CAD
TCA2478-25G TCA2478-5G
(Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetic acid ≥98.0%
(Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetic acid

CAS Number: 65872-41-5
MDL Number: MFCD00010416
Molecular Formula: C6H7N3O3S
Molecular Weight: 201.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 204
Lambda max.: 273 nm (MeOH)


Other Product Names / Synonyms: syn-Thiazoximic Acid

Formula: C₆H₇N₃O₃S
Melting Pt: 204 °C
MDL Number: MFCD00010416
CAS Number: 65872-41-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2254985573309ca7cdd772f2f36fa157a65ca704&vid=2254985573309ca7cdd772f2f36fa157a65ca704&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR