(-)-Ambroxide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ambroxide, (3aR,5aS,9aS,9bR)-3a,6,6,9a-Tetramethyldodecahydronaphtho[2,1-b]furan, 1,5,5,9-Tetramethyl-13-oxatricyclo[8.3.0.0(4,9)]tridecane

A2472-25G A2472-5G
TCA2472-25GEA 328.66 CAD
TCA2472-25G TCA2472-5G
(-)-Ambroxide ≥98.0% (by GC)
(-)-Ambroxide

CAS Number: 6790-58-5
MDL Number: MFCD00134491
Molecular Formula: C16H28O
Molecular Weight: 236.40
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 77
Specific rotation [a]20/D: -25 deg (C=1, CHCl3)


Other Product Names / Synonyms: 1,5,5,9-Tetramethyl-13-oxatricyclo[8.3.0.0(4,9)]tridecane

Formula: C₁₆H₂₈O
MW: 236.40 g/mol
Melting Pt: 77 °C
MDL Number: MFCD00134491
CAS Number: 6790-58-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a01125a1236cf1407040f68ef43973166d16575&vid=2a01125a1236cf1407040f68ef43973166d16575&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR