3-Fluoro-4-(trifluoromethyl)aniline ≥98.0%
Supplier: TCI America
Synonyms:
3-Fluoro-4-trifluoromethylaniline, 4-Amino-alpha,alpha,alpha,2-tetrafluorotoluene, α,α,α,3-Tetrafluoro-p-toluidine, 4-Amino-2-fluorobenzotrifluoride, 4-Amino-α,α,α,2-tetrafluorotoluene
CAS Number: 69411-68-3
MDL Number: MFCD00190120
Molecular Formula: C7H5F4N
Molecular Weight: 179.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 56
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Fluoro-4-trifluoromethylaniline
Formula:
C₇H₅F₄N MW: 179.12 g/mol Melting Pt: 56 °C |
MDL Number:
MFCD00190120 CAS Number: 69411-68-3 UN: 2811 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...