2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole ≥98.0% (by HPLC)

Supplier: TCI America

A2462-25G A2462-5G
TCA2462-25GEA 379.2 CAD
TCA2462-25G TCA2462-5G
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole ≥98.0% (by HPLC)
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole

CAS Number: 10444-89-0
MDL Number: MFCD00003109
Molecular Formula: C3H2F3N3S
Molecular Weight: 169.13
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 223

Formula: C₃H₂F₃N₃S
Melting Pt: 223 °C
MDL Number: MFCD00003109
CAS Number: 10444-89-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1ce212ac74edcd66b3ca03be8a1f217d229a638&vid=b1ce212ac74edcd66b3ca03be8a1f217d229a638&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR