2-Amino-6-(trifluoromethyl)benzothiazole ≥98.0%

Supplier: TCI America

Synonyms: 6-(Trifluoromethyl)benzo[d]thiazol-2-amine

A2461-1G A2461-5G
TCA2461-1GEA 74.26 CAD
TCA2461-1G TCA2461-5G
2-Amino-6-(trifluoromethyl)benzothiazole ≥98.0%
2-Amino-6-(trifluoromethyl)benzothiazole

CAS Number: 777-12-8
MDL Number: MFCD00269597
Molecular Formula: C8H5F3N2S
Molecular Weight: 218.20
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 116

Formula: C₈H₅F₃N₂S
Melting Pt: 116 °C
MDL Number: MFCD00269597
CAS Number: 777-12-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccea8d5def19f649b86e674608ae3f86d647b60a&vid=ccea8d5def19f649b86e674608ae3f86d647b60a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR