Alendronate sodium trihydrate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

A2456-25G A2456-5G
TCA2456-25GEA 441.29 CAD
TCA2456-25G TCA2456-5G
Alendronate sodium trihydrate ≥97.0% (by titrimetric analysis)
Alendronate sodium trihydrate

CAS Number: 121268-17-5
MDL Number: MFCD01748233
Molecular Formula: C4H13NO7P2
Molecular Weight: 271.08
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Storage Temperature: <0°C


Product Categories: Bioscience, Anti-osteoporosis Ingredients
Other Product Names / Synonyms: Alendronic Acid Monosodium Salt, (4-Amino-1-hydroxy-1-phosphonobutyl)phosphonic Acid Monosodium Salt, Monosodium (4-Amino-1-hydroxy-1-phosphonobutyl)phosphonate

Formula: C₄H₁₂NaNO₇P₂·3H₂O
MDL Number: MFCD01748233
CAS Number: 121268-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c39f487e322fc19ca19dce8507abd3748d219f5d&vid=c39f487e322fc19ca19dce8507abd3748d219f5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR