2-Amino-4-methoxybenzothiazole ≥98.0%

Supplier: TCI America

Synonyms: 4-Methoxybenzothiazol-2-ylamine

A2454-1G A2454-5G
TCA2454-1GEA 127.98 CAD
TCA2454-1G TCA2454-5G
2-Amino-4-methoxybenzothiazole ≥98.0%
2-Amino-4-methoxybenzothiazole

CAS Number: 5464-79-9
MDL Number: MFCD00005792
Molecular Formula: C8H8N2OS
Molecular Weight: 180.23
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 156

Formula: C₈H₈N₂OS
Melting Pt: 156 °C
MDL Number: MFCD00005792
CAS Number: 5464-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c5a14ee04d66c7fc415b83c30b52683c5ce3d21&vid=5c5a14ee04d66c7fc415b83c30b52683c5ce3d21&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR