(3-Acetamidophenyl)boronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
A2453-1G
A2453-5G
TCA2453-1GEA
154.84
CAD
TCA2453-1G
TCA2453-5G
(3-Acetamidophenyl)boronic acid (contains varying amounts of Anhydride)
(3-Acetamidophenyl)boronic acid
CAS Number: 78887-39-5
MDL Number: MFCD00236013
Molecular Formula: C8H10BNO3
Molecular Weight: 178.98
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 140
Other Product Names / Synonyms: 3-Acetamidobenzeneboronic Acid
Formula:
C₈H₁₀BNO₃ Melting Pt: 140 °C |
MDL Number:
MFCD00236013 CAS Number: 78887-39-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...