Amisulpride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-ethylsulfonyl-2-methoxybenzamide, 4-Amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-ethylsulphonyl-2-methoxybenzamide

A2450-100MG A2450-1G
TCA2450-100MGEA 235.42 CAD
TCA2450-100MG TCA2450-1G
Amisulpride ≥98.0% (by HPLC, titration analysis)
Amisulpride

CAS Number: 71675-85-9
MDL Number: MFCD00866691
Molecular Formula: C17H27N3O4S
Molecular Weight: 369.48
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 127
Storage Temperature: 0-10°C


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 4-Amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-ethylsulfonyl-2-methoxybenzamide

Formula: C₁₇H₂₇N₃O₄S
MW: 369.48 g/mol
Melting Pt: 127 °C
MDL Number: MFCD00866691
CAS Number: 71675-85-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fef5dedf23a3f90e9b462ad16efb31ed7c59d38&vid=4fef5dedf23a3f90e9b462ad16efb31ed7c59d38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR