DL-Alanine methyl ester hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: H-DL-Ala-OMe.HCl, H-DL-Ala-OMe·HCl

A2446-25G A2446-5G
TCA2446-25GEA 104.28 CAD
TCA2446-25G TCA2446-5G
DL-Alanine methyl ester hydrochloride ≥98.0%
DL-Alanine methyl ester hydrochloride

CAS Number: 13515-97-4
MDL Number: MFCD00035523
Molecular Formula: C4H9NO2
Molecular Weight: 139.58
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 158


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-DL-Ala-OMe.HCl

Formula: C₄H₉NO₂
Melting Pt: 158 °C
MDL Number: MFCD00035523
CAS Number: 13515-97-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c91f9fcb41a475d4df5a5364d4e2e48348b284b&vid=6c91f9fcb41a475d4df5a5364d4e2e48348b284b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR