3-Amino-4-bromobenzoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A2441-25G
A2441-5G
TCA2441-25GEA
549.52
CAD
TCA2441-25G
TCA2441-5G
3-Amino-4-bromobenzoic acid ≥98.0% (by HPLC, titration analysis)
3-Amino-4-bromobenzoic acid
CAS Number: 2840-29-1
MDL Number: MFCD00579106
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 228
Formula:
C₇H₆BrNO₂ Melting Pt: 228 °C |
MDL Number:
MFCD00579106 CAS Number: 2840-29-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...