4-Amino-6-hydroxypyrimidine ≥98.0%

Supplier: TCI America

A2439-1G A2439-5G
TCA2439-1GEA 52.93 CAD
TCA2439-1G TCA2439-5G
4-Amino-6-hydroxypyrimidine ≥98.0%
4-Amino-6-hydroxypyrimidine

CAS Number: 1193-22-2
MDL Number: MFCD00051502
Molecular Formula: C4H5N3O
Molecular Weight: 111.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 265


Other Product Names / Synonyms: 6-Amino-4-pyrimidinol, 6-Amino-4(1H)-pyrimidinone

Formula: C₄H₅N₃O
MW: 111.10 g/mol
Melting Pt: 265 °C
MDL Number: MFCD00051502
CAS Number: 1193-22-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=991af1fbe1e1be4908216dbbc520f4752a886133&vid=991af1fbe1e1be4908216dbbc520f4752a886133&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR