5-Amino-3-phenylisoxazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

A2437-1G A2437-5G
TCA2437-1GEA 142.2 CAD
TCA2437-1G TCA2437-5G
5-Amino-3-phenylisoxazole ≥98.0% (by HPLC, total nitrogen)
5-Amino-3-phenylisoxazole

CAS Number: 4369-55-5
MDL Number: MFCD00464114
Molecular Formula: C9H8N2O
Molecular Weight: 160.18
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 113

Formula: C₉H₇NO
Melting Pt: 113 °C
MDL Number: MFCD00464114
CAS Number: 4369-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab709fd3eb86e8d15f53cd07b257b7e9896cd170&vid=ab709fd3eb86e8d15f53cd07b257b7e9896cd170&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR