N-(2-Aminoethyl)maleimide hydrochloride ≥93.0%

Supplier: TCI America

Synonyms: N-(2-Aminoethyl)maleimide HCl, N-(2-Aminoethyl)maleimide Hydrochloride, 2-Maleimidoethylamine hydrochloride

A2436-1G A2436-200MG
TCA2436-1GEA 753.66 CAD
TCA2436-1G TCA2436-200MG
N-(2-Aminoethyl)maleimide hydrochloride ≥93.0%
N-(2-Aminoethyl)maleimide hydrochloride

CAS Number: 134272-64-3
MDL Number: MFCD09800491
Molecular Formula: C6H8N2O2
Molecular Weight: 176.60
Purity/Analysis Method: >93.0% (HPLC,N)
Form: Crystal
Melting point (°C): 163

Formula: C₆H₈N₂O₂
Melting Pt: 163 °C
MDL Number: MFCD09800491
CAS Number: 134272-64-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b5111396fe2bf3f9a911331c485ee177bc6d728&vid=5b5111396fe2bf3f9a911331c485ee177bc6d728&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR