3-Amino-4-hydroxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2432-1G A2432-5G
TCA2432-1GEA 36.34 CAD
TCA2432-1G TCA2432-5G
3-Amino-4-hydroxypyridine ≥98.0% (by GC, titration analysis)
3-Amino-4-hydroxypyridine

CAS Number: 6320-39-4
MDL Number: MFCD04114230
Molecular Formula: C5H6N2O
Molecular Weight: 110.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 150


Other Product Names / Synonyms: 3-Amino-4-pyridinol

Formula: C₅H₆N₂O
Melting Pt: 150 °C
MDL Number: MFCD04114230
CAS Number: 6320-39-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7653560338524b63011a39586e555a9e9cdb22d&vid=28004ddc297b6e04e65e10c45ab966f615f93391&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR