Amezinium methyl sulfate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-Amino-6-methoxy-1-phenylpyridazinium Methyl sulfate, 4-Amino-6-methoxy-1-phenylpyridazinium Methyl Sulfate, Amezinium Methyl sulphate, 4-Amino-6-methoxy-1-phenylpyridazin-1-ium methyl sulfate, Lu-1631, 4-Amino-6-methoxy-1-phenylpyridazinium Methyl Sulphate, Amezinium methylsulphate, Amezinium methylsulfate, Amezinium metilsulfate, 4-Amino-6-methoxy-1-phenylpyridazin-1-ium methyl sulphate, Amezinium Methyl sulfate

A2425-1G A2425-5G
TCA2425-1GEA 59.57 CAD
TCA2425-1G TCA2425-5G
Amezinium methyl sulfate ≥98.0% (by HPLC)
Amezinium methyl sulfate

CAS Number: 30578-37-1
Molecular Formula: C12H15N3O5S
Molecular Weight: 313.33
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 176


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 4-Amino-6-methoxy-1-phenylpyridazinium Methyl Sulfate

Formula: C₁₂H₁₅N₃O₅S
MW: 313.33 g/mol
Melting Pt: 176 °C
MDL Number: MFCD00865972
CAS Number: 30578-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5488ff45958370e3154f19641eee4b6cfc80bc7f&vid=5488ff45958370e3154f19641eee4b6cfc80bc7f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR