5-Acetyl-2-bromopyridine ≥98.0%

Supplier: TCI America

Synonyms: 3-Acetyl-6-bromopyridine

A2421-1G A2421-5G
TCA2421-1GEA 72.68 CAD
TCA2421-1G TCA2421-5G
5-Acetyl-2-bromopyridine ≥98.0%
5-Acetyl-2-bromopyridine

CAS Number: 139042-59-4
MDL Number: MFCD04974527
Molecular Formula: C7H6BrNO
Molecular Weight: 200.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 126

Formula: C₇H₆BrNO
MW: 200.03 g/mol
Melting Pt: 126 °C
MDL Number: MFCD04974527
CAS Number: 139042-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f511b72c61398de8fc1da8e5ff7eb514c578107a&vid=f511b72c61398de8fc1da8e5ff7eb514c578107a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR