3-Aminopiperidin-2-one ≥98.0% (by GC)

Supplier: TCI America

A2417-1G A2417-5G
TCA2417-1GEA 276.78 CAD
TCA2417-1G TCA2417-5G
3-Aminopiperidin-2-one ≥98.0% (by GC)
3-Aminopiperidin-2-one

CAS Number: 1892-22-4
MDL Number: MFCD06809610
Molecular Formula: C5H10N2O
Molecular Weight: 114.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Storage Temperature: >-20°C


Other Product Names / Synonyms: 2-Amino-delta-Valerolactam

Formula: C5H10N2O
MDL Number: MFCD06809610
CAS Number: 1892-22-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cbdcce61b564459dfbfc3d515d90946e27fd1c9&vid=0cbdcce61b564459dfbfc3d515d90946e27fd1c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR